missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement C4d Antibody (C4D/9213R), FITC, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-24022F
This item is not returnable.
View return policy
Description
Complement C4d Monoclonal antibody specifically detects Complement C4d in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)
Specifications
| Complement C4d | |
| Monoclonal | |
| FITC | |
| C4, C4A, C4A2, C4A3, C4A4, C4A6, C4S, CO4, complement C4A, CPAMD2, RG | |
| Recombinant full-length human Complement C4d protein | |
| 0.1 mL | |
| Immunology, Innate Immunity | |
| 720 | |
| Store at 4°C in the dark. | |
| IgG κ |
| ELISA, Immunohistochemistry (Paraffin) | |
| C4D/9213R | |
| PBS | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction