missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aquaporin-2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 26 publications
Brand: Novus Biologicals NB110-74682
This item is not returnable.
View return policy
Description
Aquaporin-2 Polyclonal specifically detects Aquaporin-2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| Aquaporin-2 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| ADH water channel, AQP-2, AQP-CD, aquaporin 2 (collecting duct), aquaporin-2, aquaporin-CD, Collecting duct water channel protein, MGC34501, Water channel protein for renal collecting duct, water-channel aquaporin 2, WCH-CD | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, rat and mouse. 100% sequence identity with sheep, porcine, and bovine proteins. | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/ml | |
| Western Blot 1 - 2 ug/ml, Immunohistochemistry 1:100-1:200, Immunocytochemistry/Immunofluorescence 1 ug/ml, Immunohistochemistry-Paraffin 1:100-1:200, Immunohistochemistry-Frozen reported in scientific literature | |
| P34080 | |
| AQP2 | |
| Synthetic peptide made to a C-terminus portion of rat Aquaporin-2 (within residues 200-300). [Swiss-Prot# P34080] | |
| 0.1 mL | |
| Plasma Membrane Markers, Signal Transduction | |
| 359 | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction