Learn More
Hecht Karl™ Automatic Burettes
Product Code. 10318731
Change view
Click to view available options
Capacity (Metric):
25 mL
50 mL
Unit Size:
Each
Product Code. Capacity (Metric) unitSize
10318731 25 mL Each
10023891 50 mL Each
2 options
Product Code. 10318731
Brand: Hecht Karl™ 40820025
Only null left
Add to basket
Add to basket
Designed with improved special pinch cock with titration screw for precise regulation. VITLab™ Automatic Burettes are standard type with 1.5 times the tolerance of Class AS.
  • Schilling burettes
  • Plastic burette clamp, chemical-resistant, allows discharge drop by drop by slightly pushing
  • Polyethylene storage bottles, plastic base
  • Burette fills by slightly pressing the storage bottle displaying automatic zero point adjustment
  • Plastic tubing as riser
  • Upper part of the burette is closed, Pellet titration burette design avoids soiling
  • Burette with Schellbach stripe and fused graduation “ASSISTENT-PERMANENT”, calibrated, faceted outlet tip
Specifications
Capacity (Metric) 25 mL
Material Soda Lime Glass
Graduations 0.1 mL
Includes Plastic Filling Tube, Polyethylene Bottle with Plastic Base Burette
Tolerance ±0.1 mL
Type Automatic Buret