missing translation for 'onlineSavingsMsg'
Learn More

Arachidonic Acid Methyl Ester, TRC

Product Code. 30137746
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137746 100 mg 100mg
30010843 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30137746 Supplier Toronto Research Chemicals Supplier No. TRCA765005100MG
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2566-89-4
Molecular Formula C21 H34 O2
Molecular Weight (g/mol) 318.49
Synonym 5,8,11,14-Eicosatetraenoic acid, methyl ester, (all-Z)- (8CI), Arachidonic acid, methyl ester (6CI,7CI), Methyl 5Z,8Z,11Z,14Z-eicosatetraenoate, Methyl all-cis-5,8,11,14-eicosatetraenoate, Methyl arachidonate, Methyl cis,cis,cis,cis-eicosa-5,8,11,14-tetraenoate
IUPAC Name methyl (5Z,8Z,11Z,14Z)-icosa-5,8,11,14-tetraenoate
SMILES CCCCC\C=C/C\C=C/C\C=C/C\C=C/CCCC(=O)OC

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C21H34O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21(22)23-2/h7-8,10-11,13-14,16-17H,3-6,9,12,15,18-20H2,1-2H3/b8-7-,11-10-,14-13-,17-16-
Formula Weight 318.2559
Percent Purity >95%
Chemical Name or Material Arachidonic acid methyl ester
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.