Learn More
Invitrogen™ ARC Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA14514
Description
PA1-4514 detects splice variants Nop30/ARC of the nucleolar protein Nop30 in human samples. PA1-4514 has been used successfully in Western blot procedures. By Western blot this antibody detects a ~25 kDa protein representing recombinant Nop30. The PA1-4514 immunogen corresponds to amino acid residues 125-141 of the 25 kDa human Nop30/ARC protein.
ARC (also known as NOL3 - Nucleolar protein 3) has two isoforms. Isoform 1 is involved in RNA splicing. Isoform 2 functions as an apoptosis repressor that blocks multiple modes of cell death. ARC inhibits extrinsic apoptotic pathways two different ways. The first is by interacting with FAS and FADD upon FAS activation blocking death-inducing signaling complex (DISC) assembly. The second way is by interacting with CASP8 in a mitochondria localization-and phosphorylation-dependent manner, which in turn limits the amount of soluble CASP8 avaliable for DISC-mediated activation. ARC also inhibits intrinsic apoptotic pathways in response to a wide range of stresses through interactions with BAX. This results in BAX inactivation and prevents mitochondrial dysfunction and release of pro-apoptotic factors. Mutations affecting the gene can results in myoclonus, familiam cortical.
Specifications
| ARC | |
| Polyclonal | |
| Unconjugated | |
| NOL3 | |
| activity regulated cytoskeletal-associated protein; activity regulated cytoskeleton associated protein; activity-regulated cytoskeleton-associated protein; Activity-regulated gene 3.1 protein; activity-regulated gene 3.1 protein homolog; Apoptosis repressor with CARD; ARC; arc {ECO:0000312; ARC/ARG3.1; Arc3.1; Arg3.1; B430311C09Rik; C86064; FCM; hArc; KIAA0278; mArc; Muscle-enriched cytoplasmic protein; MYC; MYP; Nol3; NOP; Nop 30; Nop30; Nucleolar protein 3; nucleolar protein 3 (apoptosis repressor with CARD domain); nucleolar protein of 30 kDa; rg3.1; RGD:62037} | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8996 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| O60936 | |
| NOL3 | |
| Synthetic peptide: P(125) E L Q T L T R P G A L R A P R R(141). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.