Learn More
Thermo Scientific™ Arctic Express™ Dual Storage Systems
Tous les produits Thermo Scientific ProduitsDescription
Protect nonhazardous samples during shipment and storage using Thermo Scientific™ Arctic Express™ Storage Systems. Featuring excellent flexibility and safety features, these systems can be used as dry shippers or laboratory cryo vessels.
Features:
- When filled with liquid nitrogen and placed in the laboratory, unit can be used as a 'can and cane' storage system indefinitely with static holding times of up to 125 days.
- Aluminum vessels come with lockable lid and hold six numbered, color-coded, stainless-steel cans (included), which accommodate canes that hold ampules.
Specifications
Specifications
| Type | Manual Fill LN2 Dewar |
| Capacity (Metric) | 28 L |
| No. of Doors | 1 |
| Model | Arctic Express Dual 28 |
| Static Holding Time | 21 shipper/50 lab |
| Diameter (English) | 18.2 in. |
| Diameter (English) Inside Neck | 3.8 in. |
| Diameter (Metric) | 46.2 cm |
| Diameter (Metric) Inside Neck | 9.7 cm |
| Vial Capacity | 630 x 2mL |