Learn More
ARFGAP1 Monoclonal Antibody (OTI1H4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502594S
Description
ARFGAP1 Monoclonal antibody specifically detects ARFGAP1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
| ARFGAP1 | |
| Monoclonal | |
| 0.7 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8N6T3 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 55738 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI1H4 | |
| Unconjugated | |
| ARFGAP1 | |
| ADP ribosylation factor GTPase activating protein 1; ADP-ribosylation factor 1 GTPase activating protein; ADP-ribosylation factor 1 GTPase-activating protein; ADP-ribosylation factor GTPase activating protein 1; ADP-ribosylation factor GTPase-activating protein 1; AI115377; ARF GAP 1; Arf GAP1; ARF1 GAP; ARF1-directed GTPase-activating protein; Arf1gap; ARFGAP1; EGK_02194; GAP protein; HRIHFB2281; RP11-261N11.1 | |
| Full length human recombinant protein of human ARFGAP1 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.