missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Argininosuccinate Lyase Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
379.00€ - 549.00€
Specifications
| Antigen | Argininosuccinate Lyase |
|---|---|
| Dilution | Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000-1:2500 |
| Applications | Immunohistochemistry (Paraffin), Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18482502
|
Novus Biologicals
NBP1-87462-25ul |
25 μL |
379.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18256687
|
Novus Biologicals
NBP1-87462 |
0.1 mL |
549.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Argininosuccinate Lyase Polyclonal specifically detects Argininosuccinate Lyase in Human, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Argininosuccinate Lyase | |
| Immunohistochemistry (Paraffin), Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| Human, Canine | |
| argininosuccinase, argininosuccinate lyase, Arginosuccinase, ASAL, EC 4.3.2.1 | |
| ASL | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000-1:2500 | |
| Polyclonal | |
| Rabbit | |
| metabolism | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 435 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:MASESGKLWGGRFVGAVDPIMEKFNASIAYDRHLWEVDVQGSKAYSRGLEKAGLLTKAEMDQILHGLDKVAEEWAQGTFKLNSNDED | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title