missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARHGAP11B Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-06142CL1
This item is not returnable.
View return policy
Description
ARHGAP11B Polyclonal antibody specifically detects ARHGAP11B in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| ARHGAP11B | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| CoraFluor 1 | |
| B'-T, FAM7B1, Family With Sequence Similarity 7, Member B1, GAP (1-8), Rho GTPase Activating Protein 11B, Rho GTPase-Activating Protein 11B, Rho-Type GTPase-Activating Protein 11B | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human ARHGAP11B. | |
| 0.1 mL | |
| GPCR, Signal Transduction | |
| 89839 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction