missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARHGAP24 Rabbit anti-Canine, Human, Mouse, Rat, Zebrafish, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18834-1-AP-150UL
This item is not returnable.
View return policy
Description
The antibody may recognize all the isoforms of ARHGAP24 except isoform 5.
Rho GTPase-activating protein involved in cell polarity, cell morphology and cytoskeletal organization. Acts as a GTPase activator for the Rac-type GTPase by converting it to an inactive GDP-bound state. Controls actin remodeling by inactivating Rac downstream of Rho leading to suppress leading edge protrusion and promotes cell retraction to achieve cellular polarity. Able to suppress RAC1 and CDC42 activity in vitro. Overexpression induces cell rounding with partial or complete disruption of actin stress fibers and formation of membrane ruffles, lamellipodia, and filopodia. Isoform 2 is a vascular cell-specific GAP involved in modulation of angiogenesis.Specifications
| ARHGAP24 | |
| Polyclonal | |
| Unconjugated | |
| ARHGAP24 | |
| ARHGAP24, Filamin A associated RhoGAP, FILGAP, p73, p73RhoGAP, RC GAP72, RCGAP72, Rho type GTPase activating protein 24, RhoGAP of 73 kDa, Sarcoma antigen NY SAR 88 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 100005116, 231532, 305156, 478463, 83478 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.31 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5U2Z7, Q8C4V1, Q8N264 | |
| ARHGAP24 | |
| Peptide | |
| 150 μL | |
| Primary | |
| Canine, Human, Mouse, Rat, Zebrafish | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction