missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARHGAP25 Antibody (OTI2D5), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71508G
This item is not returnable.
View return policy
Description
ARHGAP25 Monoclonal specifically detects ARHGAP25 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| ARHGAP25 | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ARHGAP25 | |
| Full length recombinant protein of human ARHGAP25 (NP_055697) produced in HEK293T cells. | |
| 0.1 mL | |
| 9938.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI2D5 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| KAIA0053, KIAA0053, Rho GTPase activating protein 25, rho GTPase-activating protein 25, Rho-type GTPase-activating protein 25 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction