Learn More
ARHGAP25 Monoclonal Antibody (OTI12E6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501744S
Description
ARHGAP25 Monoclonal antibody specifically detects ARHGAP25 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
| ARHGAP25 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P42331 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 9938 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI12E6 | |
| Unconjugated | |
| ARHGAP25 | |
| A130039I20Rik; ARHGAP25; epididymis secretory protein Li 308; HEL-S-308; KAIA0053; KIAA0053; RGD1562105; Rho GTPase activating protein 25; rho GTPase-activating protein 25; Rho-type GTPase-activating protein 25 | |
| Full length human recombinant protein of human ARHGAP25 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.