missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARHGDIB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16122-1-AP-150UL
This item is not returnable.
View return policy
Description
Acts as a ventralizing factor during embryogenesis. Inhibits axin-mediated JNK activation by binding axin and disrupting axin homodimerization. This in turn antagonizes a Wnt/beta-catenin-independent dorsalization pathway activated by AXIN/JNK-signaling.Specifications
| ARHGDIB | |
| Polyclonal | |
| Unconjugated | |
| ARHGDIB | |
| ARHGDIB, D4, GDIA2, GDID4, Ly GDI, LYGDI, RAP1GN1, Rho GDI 2, Rho GDI beta, RhoGDI2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11857, 362456, 397 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.14 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P52566, Q61599 | |
| Arhgdib | |
| ARHGDIB Fusion Protein Ag9078 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction