Learn More
Invitrogen™ ARHGEF11 Recombinant Rabbit Monoclonal Antibody (1I7H7)

Rabbit Recombinant Monoclonal Antibody
176.00€ - 436.00€
Specifications
| Antigen | ARHGEF11 |
|---|---|
| Clone | 1I7H7 |
| Concentration | 0.97 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA, Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30282774
|
Invitrogen™
MA557913 |
20 μL |
176.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30283352
![]() |
Invitrogen™
MA557914 |
100 μL |
436.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Immunogen sequence: QGVDQSPKPL IIGPEEDYDP GYFNNESDII FQDLEKLKSR PAHLGVFLRY IFSQADPSPL LFYLCAEVYQ QASPKDSRSL GKDIWNIFLE KNAPLRVKIP EMLQAEIDSR LRNSEDARGV LCEAQEAAMP EIQEQIHDYR TKRTLGLGSL YGENDLLDLD GDPLRERQVA EKQLAALGDI LSKYEEDRSA PMDFALNTYM SHAGIRLREA.
ARHGEF11 is a 1527 amino acid protein with tandem dbl homology and pleckstrin homology domains, PDZ domain, two proline rich sequences and a regulatory G protein-signaling (RGS) sequence. It acts as guanine nucleotide exchange factor (GEF) for RhoA GTPase, but not for RAC1 or CDC42 and acts as GTPase-activating protein (GAP) for GNA12 and GNA13. It forms a complex with G proteins and stimulates neurite through retraction Rho-dependent signals. ARHGEF11 increases glutamate transport 4-fold. It interacts with EAAT4 and modulates its uptake activity and perisynaptic distribution at glutamatergic synapses. It is known to induce the reorganization of the actin cytoskeleton and its overexpression induces the formation of membrane ruffling and filopodia. It is ubiquitously expressed, in brain, lung, liver, kidney and skeletal muscle.Specifications
| ARHGEF11 | |
| 0.97 mg/mL | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| Arhgef11; B930073M02; E130307F09; glutamate transporter EAAT4 associated protein 48; glutamate transporter EAAT4-associated protein 48; Gtrap48; KIAA0380; mKIAA0380; PDZ-RhoGEF; Prg; Rho guanine exchange factor (GEF) 11; Rho guanine nucleotide exchange factor (GEF) 11; rho guanine nucleotide exchange factor 11; RhoA-specific guanine nucleotide exchange factor; RhoGEF; RhoGEF glutamate transport modulator; RhoGEF glutamate transport modulator GTRAP48; RP11-356J7.2 | |
| ARHGEF11 | |
| Primary | |
| Protein A |
| 1I7H7 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Mouse | |
| ARHGEF11 | |
| 213498 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 281-490 of human ARHGEF11 (NP_055599.1). | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.