Learn More
Arizona Residual Solvents VOC Mixture 290 7500 μg/mL in N,N-Dimethylacetamide, Dr. Ehrenstorfer
Product Code. 30314634
Change view
Click to view available options
Quantity:
5 x 1 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30314634 5 x 1 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30314634 Supplier Dr. Ehrenstorfer Supplier No. DRES50000290DA
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
CAS 75-28-5, 106-97-8, 74-98-6
Quantity 5 x 1 mL
Certifications/Compliance ISO 17034
Solvent or Matrix N,N-Dimethylacetamide