missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARPP19 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103696
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total ARPP19.
Protein phosphatase inhibitor that specifically inhibits protein phosphatase 2A (PP2A) during mitosis. When phosphorylated at Ser-62 during mitosis, specifically interacts with PPP2R2D (PR55-delta) and inhibits its activity, leading to inactivation of PP2A, an essential condition to keep cyclin-B1-CDK1 activity high during M phase. May indirectly enhance GAP-43 expression by binding to the NGF-regulatory region of its mRNA.
Specifications
| ARPP19 | |
| Polyclonal | |
| Unconjugated | |
| ARPP19 | |
| 19kDa; 2700024H10Rik; Arpp12; ARPP16; ARPP-16; Arpp19; ARPP-19; AW559096; cAMP regulated phosphoprotein 19; cAMP regulated phosphoprotein 19kDa; cAMP-regulated phosphoprotein 19; cAMP-regulated phosphoprotein 19kDa; cAMP-regulated phosphoprotein ARPP-19; cAMP-regulated phosphoprotein, 19kDa; cyclic AMP phosphoprotein, 19 kD; cyclic AMP phosphoprotein, 19 kDa; cyclic AMP phosphoprotein, 19kD; ENSAL | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10776, 59046, 60336 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P56211, P56212, Q712U5 | |
| ARPP19 | |
| A synthesized peptide derived from human ARPP19(Accession P56211), corresponding to amino acid residues M1-K42. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction