Learn More
Applied Biosystems™ Array Card RNaseP Verification Kit
Product Code. 15890393 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 array
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15890393 1 array Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15890393 Supplier Applied Biosystems™ Supplier No. 4432522
  • ONLINE EXCLUSIVE PRICE
    3560.00€
    2242.80€ / Pack of 1
    Save 1317.20€
    37% Off
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The Array Card RNaseP Verification Kit is used to verify your real-time PCR instrument with Array Card Block
  • The Array Card RNaseP Verification Kit is used to verify your real-time PCR instrument with Array Card Block.
  • Confirm the performance of your instrument by running the RNaseP assay. Please see your instrument user guide for full instructions on use.
  • The Array Card RNaseP Verification Plate is preloaded with the necessary reagents to detect and quantitate genomic copies of the human RNase P gene, a single-copy gene encoding the RNA moiety of the RNase P enzyme. Each well contains preloaded reaction mix (1X TaqMan™ Universal PCR Master Mix, RNase P primers, and FAM™ dye-labeled probe) and template.
  • The verification plate must demonstrate the ability to distinguish between 5,000 and 10,000 genomic equivalents with a 99.7% confidence level for a subsequent sample run in a single well.
  • To ensure accurate results, it is recommended that the performance of your instrument be verified every six months.
TRUSTED_SUSTAINABILITY
Specifications
Calibrated Dye Proprietary Mix
For Use With (Equipment) ViiA™ 7 Dx System, QuantStudio™ Dx
Product Type Array Card RNAseP Verification Kit
For Use With (Application) Real Time PCR (qPCR)
Quantity 1 array
Calibration Type Instrument Verification Plate
PCR Method qPCR