missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Arrestin 3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
189.00€ - 549.00€
Specifications
| Antigen | Arrestin 3 |
|---|---|
| Concentration | 1 mg/mL |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18621998
|
Novus Biologicals
NBP2-41249-0.025mg |
0.025 mg |
189.00€
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18128118
|
Novus Biologicals
NBP2-41249 |
0.1 mg |
549.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Arrestin 3 Polyclonal specifically detects Arrestin 3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Arrestin 3 | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| arrestin 3, retinal (X-arrestin), arrestin 4, arrestin-C, ARRXcArr, CAR, C-arrestin, Cone arrestin, Retinal cone arrestin-3, X-arrestin | |
| ARR3 | |
| IgG | |
| Affinity Purified | |
| 47 kDa |
| 1 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Cell Biology, Cellular Markers, GPCR, Neuroscience, Neurotransmission, Sensory Systems, Signal Transduction, Virology Bacteria and Parasites, Vision | |
| PBS with 0.02% Sodium Azide | |
| 407 | |
| Antibody was raised against a 17 amino acid peptide near the carboxy terminus of human ARR3. The immunogen is located within the last 50 amino acids of ARR3. Amino Acid Squence: RKGEEESQKAVEAEGDE | |
| Primary | |
| At least three isoforms of ARR3 are known to exist. This antibody is predicted to not cross-react with other members of the arrestin protein family. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title