Learn More
Abeba™ Arrow 35600 Shoes
Product Code. 15680178
Change view
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
Unit Size:
1 pair
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15680178 36 1 pair
15690178 37 1 pair
15600188 38 1 pair
15610188 39 1 pair
15620188 40 1 pair
15630188 41 1 pair
15640188 42 1 pair
15650188 43 1 pair
15660188 44 1 pair
15670188 45 1 pair
15680188 46 1 pair
15690188 47 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Product Code. 15680178 Supplier Abeba™ Supplier No. 35600/36
Stock Up Your Lab
Use promo code 28007
View all promotions
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
Antistatic with heel patch microfiber. Sanitized™ treated.
  • CE EN ISO 20347:2012 OB A, FO SRA
  • Smooth leather, perforated
  • White
  • Slip resistance PU outsole sole
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Heel strap foldable, adjustable
  • Microfiber heel patch
  • Leather lining
Specifications
Certifications/Compliance CE EN ISO 20347:2012 OB A, FO SRA
Color White
Material Leather
Size 36
Sole Features Slip-resistant
Sole Material Polyurethane
Style 35600