missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARSA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19061-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene hydrolyzes cerebroside sulfate to cerebroside and sulfate. Defects in this gene lead to metachromatic leucodystrophy , a progressive demyelination disease which results in a variety of neurological symptoms and ultimately death. Alternatively spliced transcript variants have been described for this gene.Specifications
| ARSA | |
| Polyclonal | |
| Unconjugated | |
| ARSA | |
| ARSA, arylsulfatase A, ASA, Cerebroside sulfatase, MLD | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11883, 315222, 410 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.43 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P15289, P50428 | |
| Arsa | |
| ARSA Fusion Protein Ag13569 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction