missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Artemether
GREENER_CHOICE

Product Code. 18779576
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18779576 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18779576 Supplier Tocris Bioscience™ Supplier No. 6818/50
  • ONLINE EXCLUSIVE PRICE
    90.00€
    85.05€ / 50mg
    Save 4.95€
    5% Off
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Prevents cerebral malaria in Plasmodium berghei infected mice.

Artemether is an antimalarial. Prevents cerebral malaria in Plasmodium berghei infected mice. Exhibits synergistic effects with Methylene Blue (Cat. No. 3213) against P. falciparum in vitro. Inhibits neuroinflammation in neuronal/microglial cell co-cultures via induction of Nrf2 expression.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (3R,5aS,6R,8aS,9R,10S,12R,12aR)-Decahydro-10-methoxy-3,6,9-trimethyl-3,12-epoxy-12H-pyrano[4,3-j]-1,2-benzodioxepin
CAS 71963-77-4
Quantity 50 mg
Molecular Formula C16H26O5
Formula Weight 298.37
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.