missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Artesunate
GREENER_CHOICE

Product Code. 18747466
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18747466 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18747466 Supplier Tocris Bioscience™ Supplier No. 6827/50

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Suppresses ROS-induced activation of the NLRP3 inflammasome

Artesunate is a suppresses ROS-induced activation of the NLRP3 inflammasome. Arrests cell cycle in the G2/M phase and sensitizes human renal cell carcinoma (RCC) cell lines to sorafenib in vitro. Antimalarial.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Oxo-4-(((3R,5aS,6R,8aS,9R,10S,12R,12aR)-3,6,9-trimethyldecahydro-3H-3,12-epoxy[1,2]dioxepino[4,3-i]isochromen-10-yl)oxy)butanoic acid
CAS 88495-63-0
Quantity 50 mg
Molecular Formula C19H28O8
Formula Weight 384.42
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.