missing translation for 'onlineSavingsMsg'
Learn More

Artesunate, TRC

Product Code. 30135168
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135168 100 mg 100mg
30134065 50 mg 50mg
17984615 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135168 Supplier Toronto Research Chemicals Supplier No. TRCA777800100MG
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88495-63-0
Molecular Formula C19 H28 O8
Molecular Weight (g/mol) 384.42
Synonym 1-[(3R,5aS,6R,8aS,9R,10S,12R,12aR)-decahydro-3,6,9-trimethyl-3,12-epoxy-12H-pyrano[4,3-j]-1,2-benzodioxepin-10-yl] ester, Butanedioic acid, mono(decahydro-3,6,9-trimethyl-3,12-epoxy-12H-pyrano[4,3-j]-1,2-benzodioxepin-10-yl) ester, [3R-(3α,5aβ,6β,8aβ,9α,10β,12β,12aR*)]-, Butanedioic acid, mono[(3R,5aS,6R,8aS,9R,10S,12R,12aR)-decahydro-3,6,9-trimethyl-3,12-epoxy-12H-pyrano[4,3-j]-1,2-benzodioxepin-10-yl] ester (9CI), 3,12-Epoxy-12H-pyrano[4,3-j]-1,2-benzodioxepin, butanedioic acid deriv., Arinate, Armax 200, Arsuamoon, Arsumax, Artecan, Artesunata, Artesunate, Artesunic acid, Asumax, Cosunate, Dihydroqinghasu hemsuccinate, Gsunate Forte, NSC 712571, Plasmotrim, Plasmotrin, Qinghaozhi, Saphnate, WR 256283, Zysunate, α-Artesunic acid
IUPAC Name 4-oxo-4-[[(4S,5R,8S,9R,10S,12R,13R)-1,5,9-trimethyl-11,14,15,16-tetraoxatetracyclo[10.3.1.04,13.08,13]hexadecan-10-yl]oxy]butanoic acid
SMILES C[C@@H]1CC[C@H]2[C@@H](C)[C@@H](OC(=O)CCC(=O)O)O[C@@H]3O[C@@]4(C)CC[C@@H]1[C@@]23OO4

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C19H28O8/c1-10-4-5-13-11(2)16(23-15(22)7-6-14(20)21)24-17-19(13)12(10)8-9-18(3,25-17)26-27-19/h10-13,16-17H,4-9H2,1-3H3,(H,20,21)/t10-,11-,12+,13+,16+,17-,18-,19-/m1/s1
Formula Weight 384.1784
Chemical Name or Material Artesunate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.