missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARX Antibody (4H8), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00170302-M04
This item is not returnable.
View return policy
Description
ARX Monoclonal antibody specifically detects ARX in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| ARX | |
| Monoclonal | |
| Unconjugated | |
| NP_620689 | |
| Mouse | |
| IgG purified | |
| RUO | |
| 170302 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2b κ |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 4H8 | |
| In 1x PBS, pH 7.4 | |
| aristaless related homeobox, Aristaless-related homeobox, aristaless-related homeobox, X-linked, cancer/testis antigen 121, CT121, EIEE1, homeobox protein ARX, ISSXmental retardation, X-linked 36, mental retardation, X-linked 29, mental retardation, X-linked 32, mental retardation, X-linked 33, mental retardation, X-linked 38, mental retardation, X-linked 43, mental retardation, X-linked 54, mental retardation, X-linked 76, mental retardation, X-linked 87, MRX29, MRX32, MRX33, MRX36, MRX38, MRX43, MRX54, MRX76, MRX87, MRXS1, PRTS | |
| ARX (NP_620689, 1 a.a. ∽ 95 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MSNQYQEEGCSERPECKSKSPTLLSSYCIDSILGRRSPCKMRLLGAAQSLPAPLTSRADPEKAVQGSPKSSSAPFEAELHLPPKLRRLYGPGGGR | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction