missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASAH1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76933-0.025mg
This item is not returnable.
View return policy
Description
ASAH1 Polyclonal specifically detects ASAH1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| ASAH1 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| ACDase, ACEC 3.5.1.23, Acid CDase, acid ceramidase, Acylsphingosine deacylase, ASAH, FLJ21558, FLJ22079, N-acylsphingosine amidohydrolase, N-acylsphingosine amidohydrolase (acid ceramidase) 1, PHP, PHP32N-acylsphingosine amidohydrolase (acid ceramidase), Putative 32 kDa heart protein | |
| Rabbit | |
| 49 kDa | |
| 0.025 mg | |
| Primary | |
| Multiple isoforms of ASAH1 are known to exist. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1 -2 μg/mL ELISA 1:100-1:2000, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 2.5 μg/mL | |
| EAW63795 | |
| ASAH1 | |
| Antibody was raised against a 16 amino acid synthetic peptide near the carboxy terminus of the human ASAH1. The immunogen is located within amino acids 240 - 290 of ASAH1. Amino Acid Squence: LENSTSYEEAKNLLTK | |
| Affinity Purified | |
| RUO | |
| 427 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction