missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASCC3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17627-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a protein that belongs to a family of helicases that are involved in the ATP-dependent unwinding of nucleic acid duplexes. The encoded protein is the largest subunit of the activating signal cointegrator 1 complex that is involved in DNA repair and resistance to alkylation damage. Alternate splicing results in multiple transcript variants.Specifications
| ASCC3 | |
| Polyclonal | |
| Unconjugated | |
| ASCC3 | |
| ASC 1 complex subunit p200, ASC1p200, ASCC3, HELIC1, Helicase, ATP binding 1, RNAH, Trip4 complex subunit p200 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10973, 309887, 77987 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.41 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| E9PZJ8, F1LPQ2, Q8N3C0 | |
| Ascc3 | |
| ASCC3 Fusion Protein Ag11848 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction