missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASCL1 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
129.84€ - 351.27€
Specifications
| Antigen | ASCL1 |
|---|---|
| Concentration | 800 μg/mL |
| Applications | Western Blot, Immunohistochemistry, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
The ASCL1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ASCL1. This antibody recognizes human antigen. The ASCL1 antibody has been validated for the following applications: WB, IHC, ELISA analysis.Specifications
| ASCL1 | |
| Western Blot, Immunohistochemistry, ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| ASH1; HASH1; MASH1; bHLHa46 | |
| IgG | |
| Antigen Affinity purified | |
| Polyclonal Antibody |
| 800 μg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 429 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title