missing translation for 'onlineSavingsMsg'
Learn More

Ascorbyl Tetraisopalmitate, TRC

Product Code. 17994415
Change view
Click to view available options
Quantity:
1 g
2.5 g
5 g
Unit Size:
1g
2.5g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17994415 5 g 5g
17902165 2.5 g 2.5g
17971275 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17994415 Supplier Toronto Research Chemicals Supplier No. TRCA7870235G
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 183476-82-6
Molecular Formula C70H128O10
Molecular Weight (g/mol) 1129.76
Synonym L-Ascorbic acid 2,3,5,6-tetrakis(2-hexyldecanoate), L-Ascorbic acid tetrakis(2-hexyldecanoate), Ascorbyl tetra-2-hexyldecanoate, BV-OSC, Nikkol VC-IP, VC-IP, Vitamin C tetra-isopalmitate
IUPAC Name [(2S)-2-[(2R)-3,4-bis(2-hexyldecanoyloxy)-5-oxo-2H-furan-2-yl]-2-(2-hexyldecanoyloxy)ethyl] 2-hexyldecanoate
SMILES CCCCCCCCC(CCCCCC)C(=O)OC[C@@H]([C@@H]1C(=C(C(=O)O1)OC(=O)C(CCCCCC)CCCCCCCC)OC(=O)C(CCCCCC)CCCCCCCC)OC(=O)C(CCCCCC)CCCCCCCC

Specifications

Quantity 5 g
InChI Formula InChI=1S/C70H128O10/c1-9-17-25-33-37-45-50-58(49-41-29-21-13-5)66(71)76-57-62(77-67(72)59(51-42-30-22-14-6)54-46-38-34-26-18-10-2)63-64(79-68(73)60(52-43-31-23-15-7)55-47-39-35-27-19-11-3)65(70(75)78-63)80-69(74)61(53-44-32-24-16-8)56-48-40-36-28-20-12-4/h58-63H,9-57H2,1-8H3/t58?,59?,60?,61?,62-,63+/m0/s1
Formula Weight 1128.9508
Percent Purity >95%
Chemical Name or Material Ascorbyl Tetraisopalmitate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.