Learn More
ASIC1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 27235-1-AP-150UL
 View more versions of this product
Product Code. 16875505
  • 410.00€ / 150µL
Description
This gene encodes a member of the degenerin/epithelial sodium channel (DEG/ENaC) superfamily. The members of this family are amiloride-sensitive sodium channels that contain intracellular N and C termini, 2 hydrophobic transmembrane regions, and a large extracellular loop, which has many cysteine residues with conserved spacing. The member encoded by this gene is expressed in most if not all brain neurons, and it may be an ion channel subunit; however, its function as an ion channel remains unknown. Cation channel with high affinity for sodium, which is gated by extracellular protons and inhibited by the diuretic amiloride. ASIC1 is permeable for Ca(2+), Li(+) and K(+) and is expressed in dorsal root ganglia, sciatic nerves, olfactory bulb, neo and allocortical regions, dentate granule cells, pyramidal cells of CA1-CA3 subfields of the hippocampal formation, habenula, basolateral amygdaloid nuclei, and in the Purkinje and granule cells of the cerebellum. It is diffusely detected over most other regions of the basal ganglia, including thalamic nuclei, substantia nigra, striatum and globus pallidus, hypothalamus, midbrain, pons, medulla and choroid plexus. Isoform 3 is expressed only in dorsal root ganglion while isoform 1 is expressed in DRG, spinal chord, trigeminal ganglia and the trigeminal mesencephalic nucleus.
Specifications
Specifications
ASIC1
Polyclonal
Unconjugated
ASIC1
ACCN2, Acid sensing ion channel 1, ASIC, ASIC1, ASIC1A, BNaC2, Brain sodium channel 2, hBNaC2
Rabbit
Antigen Affinity Chromatography
RUO
11419, 41
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.5 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P78348, Q6NXK8
ASIC1
ASIC1 Fusion Protein Ag25941
150 μL
Primary
Human, Mouse