missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16645-1-AP-150UL
This item is not returnable.
View return policy
Description
ASL encodes a member of the lyase 1 family. The encoded protein forms a cytosolic homotetramer and primarily catalyzes the reversible hydrolytic cleavage of argininosuccinate into arginine and fumarate, an essential step in the liver in detoxifying ammonia via the urea cycle. Mutations in this gene result in the autosomal recessive disorder argininosuccinic aciduria, or argininosuccinic acid lyase deficiency. A nontranscribed pseudogene is also located on the long arm of chromosome 22. Alternatively spliced transcript variants encoding different isoforms have been described.Specifications
| ASL | |
| Polyclonal | |
| Unconjugated | |
| ASL | |
| argininosuccinate lyase, Arginosuccinase, ASAL, ASL | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 109900, 435, 59085 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P04424, P20673, Q91YI0 | |
| Asl | |
| ASL Fusion Protein Ag10007 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction