missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ASM Recombinant Rabbit Monoclonal Antibody (5T2N2)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

176.00€ - 436.00€

Specifications

Antigen ASM
Clone 5T2N2
Concentration 2.03 mg/mL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Applications ELISA, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30284232
View Documents
Invitrogen™
MA558119
20 μL
176.00€
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30282694
GREENER_CHOICE
View Documents
Invitrogen™
MA558120
100 μL
436.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Immunogen sequence: PARLHRIVPR LRDVFGWGNL TCPICKGLFT AINLGLKKEP NVARVGSVAI KLCNLLKIAP PAVCQSIVHL FEDDMVEVWR RSVLSPSEAC GLLLGSTCGH WDIFSSWNIS LPTVPKPPPK PPSPPAPGAP VSRILFLTDL HWDHDYLEGT DPDCADPLCC RRGSGLPPAS RPGAGYWGEY SKCDLPLRTL ESLLSGLGPA GPFDMVYWTG DIPAHDVWHQ TRQDQLRALT TVTALVRKFL GPVPVYPAVG NHESTPVNSF PPPFIEGNHS S.

The protein encoded by this gene is a lysosomal acid sphingomyelinase that converts sphingomyelin to ceramide. The encoded protein also has phospholipase C activity. Defects in this gene are a cause of Niemann-Pick disease type A (NPA) and Niemann-Pick disease type B (NPB). Three transcript variants encoding two different isoforms have been found for this gene.
Specifications

Specifications

ASM
2.03 mg/mL
ELISA, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
RUO
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
P17405, Q04519
20597, 308909, 6609
Recombinant protein (or fragment).
Antibody
5T2N2
-20°C, Avoid Freeze/Thaw Cycles
Recombinant Monoclonal
Liquid
IgG
Human, Mouse, Rat
Smpd1
Acid sphingomyelinase; ASM; aSMase; A-SMase; NPD; Smpd1; Sphingomyelin phosphodiesterase; sphingomyelin phosphodiesterase 1; sphingomyelin phosphodiesterase 1, acid lysosomal; Zn-SMase
Smpd1
Primary
Protein A
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.