missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASMT Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
190.00€ - 463.00€
Specifications
| Antigen | ASMT |
|---|---|
| Dilution | Western Blot 1:1000 - 1:5000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18623372
|
Novus Biologicals
NBP2-92663-0.02ml |
0.02 mL |
190.00€
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18648412
|
Novus Biologicals
NBP2-92663-0.1ml |
0.1 mL |
463.00€
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ASMT Polyclonal antibody specifically detects ASMT in Human, Mouse samples. It is validated for Western BlotSpecifications
| ASMT | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Amino Acids Drugs and other small molecules, Endocrinology, Neuroscience, Signal Transduction | |
| PBS with 50% glycerol, pH7.3. | |
| 438 | |
| IgG | |
| Affinity purified |
| Western Blot 1:1000 - 1:5000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| acetylserotonin N-methyltransferase, acetylserotonin O-methyltransferaseEC 2.1.1.4, ASMTY, HIOMTacetylserotonin methyltransferase (Y chromosome), HIOMTY, hydroxyindole O-methyltransferase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-298 of human ASMT (NP_001164510.1). MGSSEDQAYRLLNDYANGFMVSQVLFAACELGVFDLLAEAPGPLDVAAVAAGVRASAHGTELLLDICVSLKLLKVETRGGKAFYRNTELSSDYLTTVSPTSQCSMLKYMGRTSYRCWGHLADAVREGRNQYLETFGVPAEELFTAIYRSEGERLQFMQALQEVWSVNGRSVLTAFDLSVFPLMCDLGGDFFKDPLPEADLYILARVLHDWADGKCSHLLERIYHTCKPGGGILVIESLLDEDRRGPLLTQLYSLNMLVQTEGQERTPTHYHMLLSSAGFRDFQFKKTGAIYDAILARK | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title