missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ASMT Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144621
This item is not returnable.
View return policy
Description
ASMT Polyclonal Antibody for Western Blot
ASMT belongs to the methyltransferase superfamily, and is located in the pseudoautosomal region (PAR) at the end of the short arms of the X and Y chromosomes. The encoded enzyme catalyzes the final reaction in the synthesis of melatonin, and is abundant in the pineal gland.
Specifications
| ASMT | |
| Polyclonal | |
| Unconjugated | |
| ASMT | |
| acetylserotonin methyltransferase (Y chromosome); acetylserotonin N-methyltransferase; acetylserotonin O-methyltransferase; ASMT; ASMTY; Hiomt; HIOMTY; hydroxyindole O-methyltransfease; hydroxyindole O-methyltransferase; hydroxyindole-O-methyltransferase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 107626, 246281, 438 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 4.47 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300 | |
| B3GSH5, D3KU66, P46597 | |
| ASMT | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human ASMT. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction