missing translation for 'onlineSavingsMsg'
Learn More

Aspartame, Mikromol™

Product Code. 17997174
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17997174 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17997174 Supplier Mikromol Supplier No. MM0314.00
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Aspartame
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aspartame
CAS 22839-47-0
Analyte or Component Names Aspartame
Shipping Condition Room Temperature
Molecular Formula C14 H18 N2 O5
Certifications/Compliance ISO 17034
Synonym Aspartame, Aspartylphenylalanine methyl ester, Canderel, Dipeptide sweetener, E 951, Equal, Finn, L-Aspartame, L-Aspartyl-L-3-phenylalanine methyl ester, L-Aspartyl-L-phenylalanine methyl ester, L-Aspartyl-L-phenylalanyl methyl ester, Methyl aspartylphenylalanate, NutraSweet, Pal Sweet, Palsweet Diet, Sweet dipeptide, Zero-Cal, L-alpha-Aspartyl-L-phenylalanine 2-methyl ester, N-L-alpha-Aspartyl-L-phenylalanine 1-methyl ester, 3-Amino-N-(alpha-carboxyphenethyl)succinamic acid N-methyl ester stereoisomer, L-alpha-Aspartyl-L-phenylalanine methyl ester, alpha-Aspartame, alpha-L-Aspartyl-L-phenylalanine methyl ester, alpha-Sweet
InChI Formula InChI=1S/C14H18N2O5/c1-21-14(20)11(7-9-5-3-2-4-6-9)16-13(19)10(15)8-12(17)18/h2-6,10-11H,7-8,15H2,1H3,(H,16,19)(H,17,18)/t10-,11-/m0/s1
SMILES COC(=O)[C@H](Cc1ccccc1)NC(=O)[C@@H](N)CC(=O)O
IUPAC Name (3S)-3-amino-4-[[(2S)-1-methoxy-1-oxo-3-phenylpropan-2-yl]amino]-4-oxobutanoic acid
Molecular Weight (g/mol) 294.3
Quantity 500 mg
Formula Weight 294.1216 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.