missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASPHD1 Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00253982-B01P
This item is not returnable.
View return policy
Description
ASPHD1 Polyclonal antibody specifically detects ASPHD1 in Human samples. It is validated for Western Blot
Specifications
| ASPHD1 | |
| Polyclonal | |
| PBS (pH 7.4) | |
| aspartate beta-hydroxylase domain containing 1, aspartate beta-hydroxylase domain-containing protein 1, EC 1.14.11, EC 1.14.11.- | |
| LOC253982 (NP_859069.2, 1 a.a. - 390 a.a.) full-length human protein. MKEGRGSFSVERGPRKERETAQSGMWKGNSPAGSQGAAMEGTGGELGGQGNWGPEDAPGLLARASLIMLPWPLPLASSALTLLFGALTSLFLWYCYRLGSQDMQALGAGSRAGGVRGGPVGCSEAGGPSPGGPGDPGEGPRTEGLVSRRLRAYARRYSWAGMGRVRRAAQGGPGPGRGPGVLGIQRPGLLFLPDLPSAPFVPRDAQRHDVELLESSFPAILRDFGAVSWDFSGTTPPPRGWSPPLAPGCYQLLLYQAGRCQPSNCRRCPGAYRALRGLRSFMSANTFGNAGFSVLLPGARLEGRCGPTNARVRCHLGLKIPPGCELVVGGEPQCWAEGHCLLVDDSFLHTVAHNGSPEDGPRVVFIVDLWHPNVAGAERQALDFVFAPDP | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| NP_859069.2 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 253982 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction