missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Atenolol EP Impurity H (Atenolol USP Related Compound B), Mikromol™
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mg
100 mg
missing translation for 'unitSize'
100mg
25mg
Description
Atenolol EP Impurity H (Atenolol USP Related Compound B)
Specifications
Specifications
| Chemical Name or Material | 2-[4-[(2RS)-2-Hydroxy-3-[(1-methylethyl)amino]propoxy]phenyl]acetonitrile |
| CAS | 29277-73-4 |
| Analyte or Component Names | 2-[4-[(2RS)-2-Hydroxy-3-[(1-methylethyl)amino]propoxy]phenyl]acetonitrile |
| Shipping Condition | Room Temperature |
| Molecular Formula | C14 H20 N2 O2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 2-[4-[(2RS)-2-Hydroxy-3-[(1-methylethyl)amino]propoxy]phenyl]acetonitrile, Atenolol Imp. H (EP), Atenolol Related Compound B, Atenolol Impurity H |
| InChI Formula | InChI=1S/C14H20N2O2/c1-11(2)16-9-13(17)10-18-14-5-3-12(4-6-14)7-8-15/h3-6,11,13,16-17H,7,9-10H2,1-2H3 |
| SMILES | CC(C)NCC(O)COc1ccc(CC#N)cc1 |
| IUPAC Name | 2-[4-[2-hydroxy-3-(propan-2-ylamino)propoxy]phenyl]acetonitrile |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction