missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ATF3 Recombinant Rabbit Monoclonal Antibody (24GB6930)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA556975

 View more versions of this product

Product Code. 30275583

  • 514.00€ / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

ATF3 Recombinant Monoclonal Antibody for Western Blot

ATF3 (Activating transcription factor 3) is a member of the mammalian activation transcription factor/cAMP responsive element-binding (CREB) protein family of transcription factors. It encodes a protein with a calculated molecular mass of 22 kD. ATF3 represses rather than activates transcription from promoters with ATF binding elements. An alternatively spliced form of ATF3 (ATF3 delta Zip) encodes a truncated form ATF3 protein lacking the leucine zipper protein-dimerization motif and does not bind to DNA. In contrast to ATF3, ATF3 delta Zip stimulates transcription presumably by sequestering inhibitory co-factors away from the promoter. It is possible that alternative splicing of the ATF3 gene may be physiologically important in the regulation of target genes. Three transcript variants encoding two different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ATF3
Recombinant Monoclonal
0.6 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P18847
ATF3
Recombinant protein of human ATF3 (aa 1-181).
100 μL
Primary
Human
Antibody
IgG
Flow Cytometry, Western Blot
24GB6930
Unconjugated
ATF3
activating transcription factor 3; ATF 3; ATF3; cAMP-dependent transcription factor ATF-3; cyclic AMP-dependent transcription factor ATF-3; Liver regeneration factor 1; Lrf1; LRF-1; LRFI; LRG-21; MGC137608 protein; transcription factor LRG-21; zgc:55526
Rabbit
Affinity chromatography
RUO
467
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.