missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATF4 Antibody (S360A-24), Novus Biologicals™
Mouse Monoclonal Antibody
174.00€ - 601.00€
Specifications
| Antigen | ATF4 |
|---|---|
| Clone | S360A-24 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18634319
|
Novus Biologicals
NBP2-42176 |
0.1 mg |
601.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18685526
|
Novus Biologicals
NBP2-42176-0.025mg |
0.025 mg |
174.00€
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ATF4 Monoclonal antibody specifically detects ATF4 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
| ATF4 | |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Lipid and Metabolism, Unfolded Protein Response, Unfolded Protein Response (UPR) | |
| PBS (pH 7.4), 50% Glycerol | |
| Activating transcription factor 4, activating transcription factor 4 (tax-responsive enhancer element B67), cAMP-dependent transcription factor ATF-4, cAMP-responsive element-binding protein 2, CREB-2DNA-binding protein TAXREB67, cyclic AMP-dependent transcription factor ATF-4, Cyclic AMP-responsive element-binding protein 2, TaxREB67, TAXREB67CREB2cAMP response element-binding protein 2, Tax-responsive enhancer element-binding protein 67, TXREB | |
| Fusion protein amino acids 25-327 of human ATF4. 86% identical to rat, and 85% identical to mouse. <50% identity with ATF5. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| S360A-24 | |
| Western Blot 1:1000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Rat | |
| P18848, NP_001666.2 | |
| 468 | |
| IgG2a | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title