missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ATF4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA586112
This item is not returnable.
View return policy
Description
Purity is > 95% by SDS-PAGE.
This gene encodes a transcription factor that was originally identified as a widely expressed mammalian DNA binding protein that could bind a tax-responsive enhancer element in the LTR of HTLV-1. The encoded protein was also isolated and characterized as the cAMP-response element binding protein 2 (CREB-2). The protein encoded by this gene belongs to a family of DNA-binding proteins that includes the AP-1 family of transcription factors, cAMP-response element binding proteins (CREBs) and CREB-like proteins. These transcription factors share a leucine zipper region that is involved in protein-protein interactions, located C-terminal to a stretch of basic amino acids that functions as a DNA binding domain. Two alternative transcripts encoding the same protein have been described. Two pseudogenes are located on the X chromsome at q28 in a region containing a large inverted duplication.
Specifications
| ATF4 | |
| Polyclonal | |
| Unconjugated | |
| Atf4 | |
| Activating transcription factor 4; activating transcription factor 4 (tax-responsive enhancer element B67); activating transcription factor 4a; activating transcription factor 4b1 (tax-responsive enhancer element B67); activating transcription factor ATF-4; ATF 4; atf4; Atf-4; atf4 {ECO:0000312; ATF4 protein; atf4a; atf4b1; C/ATF; C/EBP-related ATF; cAMP response element-binding protein 2; cAMP-dependent transcription factor ATF-4; cAMP-responsive element-binding protein 2; CREB 2; CREB2; CREB-2; Cyclic AMP-dependent transcription factor ATF-4; cyclic AMP-responsive element-binding protein 2; DNA-binding protein TAXREB67; hypothetical protein LOC509107; OTTHUMP00000199130; rATF-4; RGD:621863}; tax-re; TaxREB67; TaxREB67 homolog; tax-responsive enhancer element B67; tax-responsive enhancer element-binding protein 67; Tax-responsive enhancer element-binding protein 67 homolog; TXREB; wu:fj01c08; wu:fk30b04; zgc:85951 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11911, 468, 79255 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P18848, Q06507, Q9ES19 | |
| Atf4 | |
| Synthetic peptide, corresponding to amino acids 200-250 of Human ATF4. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction