missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATF5 Antibody (SD2099), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-67767
This item is not returnable.
View return policy
Description
ATF5 Monoclonal specifically detects ATF5 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| ATF5 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100-1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:50-1:200 | |
| activating transcription factor 5FLJ34666, ATFXHMFN0395, cAMP-dependent transcription factor ATF-5, cyclic AMP-dependent transcription factor ATF-5, Transcription factor ATFx | |
| Rabbit | |
| Protein A purified | |
| Cell Cycle and Replication | |
| 22809 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| SD2099 | |
| Unconjugated | |
| TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide | |
| ATF5 | |
| Synthetic peptide within Human ATF5 aa 203-252/ 282. (SwissProt: Q9Y2D1 Human; SwissProt: O70191 Mouse; SwissProt: Q6P788 Rat) | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction