Learn More
ATF7IP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14699-1-AP-20UL
This item is not returnable.
View return policy
Description
ATF7IP is a multifunctional nuclear protein that associates with heterochromatin. It can act as a transcriptional coactivator or corepressor depending upon its binding partners.Specifications
| ATF7IP | |
| Polyclonal | |
| Unconjugated | |
| ATF7IP | |
| ATF7IP, MCAF, p621 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 312800, 54343, 55729 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q6VMQ6, Q7TT18 | |
| Atf7ip | |
| ATF7IP Fusion Protein Ag6341 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |