Learn More
ATG16L1 Mouse anti-Human, Unconjugated, Clone: 1G5G7, Proteintech
Mouse Monoclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | ATG16L1 |
|---|---|
| Clone | 1G5G7 |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17482433
|
Proteintech
67943-1-IG-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17472433
|
Proteintech
67943-1-IG-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The ATG16L1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ATG16L1. This antibody recognizes human, mouse, rat antigen. The ATG16L1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| ATG16L1 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Human, Rat, Mouse | |
| IgG1 |
| 1G5G7 | |
| Monoclonal | |
| Mouse | |
| 55054 | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.