Learn More
uvex Athletic D5XP Safety Glove
Shop All Uvex Products
Click to view available options
Size:
6
7
8
9
10
11
Unit Size:
10 pairs
Description
- Outer material made from HPPE, steel and polyamide/elastane
- Micro-NBR foam coating on palm and fingertips
- High cut protection (Level D)
- High mechanical resistance
- Good grip on dry and (slightly) oily/wet workpieces
- Flexible safety gloves with excellent fit
- Good tactile feel
- Safety glove with knitted cuff and reinforced thumb joint
Specifications
Specifications
| Certifications/Compliance | EN 388 (4 X 4 3 D) |
| Coating Material | Nitrile Foam |
| Color | Gray, Black |
| Hand | Ambidextrous |
| Latex Free | Yes |
| Material | HPPE, Steel Fiber, Nylon, Elastane |
| Product Type | Cut-resistant Gloves |
| Size | 6 |