missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATOH8 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92353-0.1ml
This item is not returnable.
View return policy
Description
ATOH8 Polyclonal antibody specifically detects ATOH8 in Mouse, Rat samples. It is validated for Western Blot
Specifications
| ATOH8 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| ATH6, atonal homolog 6, atonal homolog 8 (Drosophila), basic helix-loop-helix transcription factor 6, BHLHA21, bHLHa21basic helix loop helix transcription factor 6, Class A basic helix-loop-helix protein 21, FLJ14708, FLJ38730, hATH6, Helix-loop-helix protein hATH-6, protein atonal homolog 8 | |
| A synthetic peptide corresponding to a sequence within amino acids 150-250 of human ATOH8 (NP_116216.2). LRPRILLCAPPARPAPSAPPAPPAPPESTVRPAPPTRPGESSYSSISHVIYNNHQDSSASPRKRPGEATAASSEIKALQQTRRLLANARERTRVHTISAAF | |
| 0.1 mL | |
| Cell Biology, Neuroscience | |
| 84913 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction