missing translation for 'onlineSavingsMsg'
Learn More

Atorvastatin Acetonide, TRC

Product Code. 30011096
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011096 50 mg 50mg
30003867 25 mg 25mg
17929684 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011096 Supplier Toronto Research Chemicals Supplier No. TRCA79169050MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 581772-29-4
Molecular Formula C36 H39 F N2 O5
Molecular Weight (g/mol) 598.7
Synonym 1,3-Dioxane-4-acetic acid, 6-[2-[2-(4-fluorophenyl)-5-(1-methylethyl)-3-phenyl-4-[(phenylamino)carbonyl]-1H-pyrrol-1-yl]ethyl]-2,2-dimethyl-, (4R,6R)-, (4R,6R)-6-[2-[2-(4-Fluorophenyl)-5-(1-methylethyl)-3-phenyl-4-[(phenylamino)carbonyl]-1H-pyrrol-1-yl]ethyl]-2,2-dimethyl-1,3-dioxane-4-acetic acid, 2-[(4R,6R)-6-[2-[2-(4-Fluorophenyl)-5-isopropyl-3-phenyl-4-(phenylcarbamoyl)-1H-pyrrol-1-yl]ethyl]-2,2-dimethyl-1,3-dioxan-4-yl]acetic acid, Atorvastatin acetonide, Lipitor acetonide
IUPAC Name 2-[(4R,6R)-6-[2-[2-(4-fluorophenyl)-3-phenyl-4-(phenylcarbamoyl)-5-propan-2-ylpyrrol-1-yl]ethyl]-2,2-dimethyl-1,3-dioxan-4-yl]acetic acid
SMILES CC(C)c1c(C(=O)Nc2ccccc2)c(c3ccccc3)c(c4ccc(F)cc4)n1CC[C@@H]5C[C@H](CC(=O)O)OC(C)(C)O5

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C36H39FN2O5/c1-23(2)33-32(35(42)38-27-13-9-6-10-14-27)31(24-11-7-5-8-12-24)34(25-15-17-26(37)18-16-25)39(33)20-19-28-21-29(22-30(40)41)44-36(3,4)43-28/h5-18,23,28-29H,19-22H2,1-4H3,(H,38,42)(H,40,41)/t28-,29-/m1/s1
Formula Weight 598.2843
Chemical Name or Material Atorvastatin Acetonide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.