missing translation for 'onlineSavingsMsg'
Learn More

Atorvastatin Methyl Ester, TRC

Product Code. 30136522
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136522 25 mg 25mg
30004584 5 mg 5mg
30011481 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136522 Supplier Toronto Research Chemicals Supplier No. TRCA79184525MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 345891-62-5
Molecular Formula C34 H37 F N2 O5
Molecular Weight (g/mol) 572.67
Synonym 1H-Pyrrole-1-heptanoic acid, 2-(4-fluorophenyl)-β,δ-dihydroxy-5-(1-methylethyl)-3-phenyl-4-[(phenylamino)carbonyl]-, methyl ester, (βR,δR)-, Methyl (βR,δR)-2-(4-fluorophenyl)-β,δ-dihydroxy-5-(1-methylethyl)-3-phenyl-4-[(phenylamino)carbonyl]-1H-pyrrole-1-heptanoate, (βR,δR)-2-(4-Fluorophenyl)-β,δ-dihydroxy-5-(1-methylethyl)-3-phenyl-4-[(phenylamino)carbonyl]-1H-pyrrole-1-heptanoic acid methyl ester, Methyl (3R,5R)-7-(2-(4-fluorophenyl)-5-isopropyl-3-phenyl-4-(phenylcarbamoyl)-1H-pyrrol-1-yl)-3,5-dihydroxyheptanoate, Atorvastatin methyl ester
IUPAC Name methyl (3R,5R)-7-[2-(4-fluorophenyl)-3-phenyl-4-(phenylcarbamoyl)-5-propan-2-ylpyrrol-1-yl]-3,5-dihydroxyheptanoate
SMILES COC(=O)C[C@H](O)C[C@H](O)CCn1c(C(C)C)c(C(=O)Nc2ccccc2)c(c3ccccc3)c1c4ccc(F)cc4

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C34H37FN2O5/c1-22(2)32-31(34(41)36-26-12-8-5-9-13-26)30(23-10-6-4-7-11-23)33(24-14-16-25(35)17-15-24)37(32)19-18-27(38)20-28(39)21-29(40)42-3/h4-17,22,27-28,38-39H,18-21H2,1-3H3,(H,36,41)/t27-,28-/m1/s1
Formula Weight 572.2687
Percent Purity >95%
Chemical Name or Material Atorvastatin Methyl Ester
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.