missing translation for 'onlineSavingsMsg'
Learn More

Atorvastatin N-(3,5-Dihydroxy-7-heptanoic Acid)amide, TRC

Product Code. 30011889
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011889 5 mg 5mg
30011379 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30011889 Supplier Toronto Research Chemicals Supplier No. TRCA7917055MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1105067-87-5
Molecular Formula 2 C40 H47 F N3 O8 . Ca
Molecular Weight (g/mol) 1473.71
Synonym (3R,5R)-7-[[(3R,5R)-7-[2-(4-Fluorophenyl)-5-(1-methylethyl)-3-phenyl-4-[(phenylamino)carbonyl]-1H-pyrrol-1-yl]-3,5-dihydroxy-1-oxoheptyl]amino]-3,5-dihydroxyheptanoic Acid Calcium Salt (2:1), Calcium (3R,5R)-7-[[(3R,5R)-7-[2-(4-fluorophenyl)-5-(1-methylethyl)-3-phenyl-4-[(phenylamino)carbonyl]-1H-pyrrol-1-yl]-3,5-dihydroxy-1-oxoheptyl]amino]-3,5-dihydroxyheptanoate (1:2)
IUPAC Name calcium;(3R,5R)-7-[[(3R,5R)-7-[2-(4-fluorophenyl)-3-phenyl-4-(phenylcarbamoyl)-5-propan-2-ylpyrrol-1-yl]-3,5-dihydroxyheptanoyl]amino]-3,5-dihydroxyheptanoate
SMILES [Ca+2].CC(C)c1c(C(=O)Nc2ccccc2)c(c3ccccc3)c(c4ccc(F)cc4)n1CC[C@@H](O)C[C@@H](O)CC(=O)NCC[C@@H](O)C[C@@H](O)CC(=O)[O-].CC(C)c5c(C(=O)Nc6ccccc6)c(c7ccccc7)c(c8ccc(F)cc8)n5CC[C@@H](O)C[C@@H](O)CC(=O)NCC[C@@H](O)C[C@@H](O)CC(=O)[O-]

Specifications

Quantity 5 mg
InChI Formula InChI=1S/2C40H48FN3O8.Ca/c2*1-25(2)38-37(40(52)43-29-11-7-4-8-12-29)36(26-9-5-3-6-10-26)39(27-13-15-28(41)16-14-27)44(38)20-18-31(46)22-32(47)23-34(49)42-19-17-30(45)21-33(48)24-35(50)51;/h2*3-16,25,30-33,45-48H,17-24H2,1-2H3,(H,42,49)(H,43,52)(H,50,51);/q;;+2/p-2/t2*30-,31-,32-,33-;/m11./s1
Formula Weight 1472.632
Chemical Name or Material Atorvastatin N-(3,5-Dihydroxy-7-heptanoic Acid)amide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.