missing translation for 'onlineSavingsMsg'
Learn More

Atorvastatin tert-Butyl Ester, Mikromol™

Product Code. 30050228
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30050228 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30050228 Supplier Mikromol Supplier No. MM0400.350025
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Atorvastatin tert-Butyl Ester
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Atorvastatin tert-Butyl Ester
CAS 134395-00-9
Analyte or Component Names Atorvastatin tert-Butyl Ester
Shipping Condition Room Temperature
Molecular Formula C37 H43 F N2 O5
Certifications/Compliance ISO 9001
Synonym 1H-Pyrrole-1-heptanoic acid, 2-(4-fluorophenyl)-β,Δ-dihydroxy-5-(1-methylethyl)-3-phenyl-4-[(phenylamino)carbonyl]-, 1,1-dimethylethyl ester, (βR,ΔR)-, 1H-Pyrrole-1-heptanoic acid, 2-(4-fluorophenyl)-β,Δ-dihydroxy-5-(1-methylethyl)-3-phenyl-4-[(phenylamino)carbonyl]-, 1,1-dimethylethyl ester, [R-(R*,R*)]-, (3R,5R)-7-[2-(4-Fluorophenyl)-5-isopropyl-3-phenyl-4-phenylcarbamoylpyrrol-1-yl]-3,5-dihydroxyheptanoic acid tert-butyl ester, Atorvastatin tert-butyl ester, Atorvastatin Calcium Impurity N, Atorvastatin Impurity N
InChI Formula InChI=1S/C37H43FN2O5/c1-24(2)34-33(36(44)39-28-14-10-7-11-15-28)32(25-12-8-6-9-13-25)35(26-16-18-27(38)19-17-26)40(34)21-20-29(41)22-30(42)23-31(43)45-37(3,4)5/h6-19,24,29-30,41-42H,20-23H2,1-5H3,(H,39,44)/t29-,30-/m1/s1
SMILES CC(C)c1c(C(=O)Nc2ccccc2)c(c3ccccc3)c(c4ccc(F)cc4)n1CC[C@@H](O)C[C@@H](O)CC(=O)OC(C)(C)C
IUPAC Name tert-butyl (3R,5R)-7-[2-(4-fluorophenyl)-3-phenyl-4-(phenylcarbamoyl)-5-propan-2-ylpyrrol-1-yl]-3,5-dihydroxyheptanoate
Molecular Weight (g/mol) 614.75
Quantity 25 mg
Formula Weight 614.3156 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.