missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP Citrate Lyase Antibody (3A5), Novus Biologicals™
Rabbit Monoclonal Antibody
343.00€ - 533.00€
Specifications
| Antigen | ATP Citrate Lyase |
|---|---|
| Clone | 3A5 |
| Dilution | Western Blot 1:500-1:5000, Flow Cytometry 1:20-1:200, ELISA, Immunocytochemistry/Immunofluorescence 1:20-1:200 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043492
|
Novus Biologicals
NBP3-26530-50ul |
50 μL |
343.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043725
|
Novus Biologicals
NBP3-26530-100ul |
100 μL |
533.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ATP Citrate Lyase Monoclonal antibody specifically detects ATP Citrate Lyase in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, ImmunocytochemistrySpecifications
| ATP Citrate Lyase | |
| Western Blot 1:500-1:5000, Flow Cytometry 1:20-1:200, ELISA, Immunocytochemistry/Immunofluorescence 1:20-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| ACLEC 2.3.3.8, ATP citrate lyase, ATP citrate synthase, ATP-citrate (pro-S-)-lyase, ATP-citrate synthase, ATPCL, Citrate cleavage enzyme, CLATP | |
| A synthesized peptide derived from Human ATP Citrate Lyase [UniProt P53396] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 3A5 | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Phospho Specific | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 47 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title