Learn More
ATP Synthase O Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA559548
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: LEEATLSELK TVLKSFLSQG QVLKLEAKTD PSILGGMIVR IGEKYVDMSV KTKIQKLGRA MREIV Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 77%.
The protein encoded by this gene is a component of the F-type ATPase found in the mitochondrial matrix. F-type ATPases are composed of a catalytic core and a membrane proton channel. The encoded protein appears to be part of the connector linking these two components and may be involved in transmission of conformational changes or proton conductance.Specifications
ATP Synthase O | |
Polyclonal | |
Unconjugated | |
ATP5PO | |
ATP synthase peripheral stalk subunit OSCP; ATP synthase subunit O, mitochondrial; ATP synthase, H+ transporting, mitochondrial F1 complex, O subunit; ATP synthase, H+ transporting, mitochondrial F1 complex, O subunit (oligomycin sensitivity conferring protein); ATP synthase, H+ transporting, mitochondrial F1 complex, O subunit; DNA segment, Chr 12, Wayne State University 28, expressed; ATP5O; ATP5PO; ATPO; D12Wsu28e; HMC08D05; human ATP synthase OSCP subunit; oligomycin sensitivity conferral protein; oligomycin sensitivity conferral protein oscp-like protein; oligomycin sensitivity conferring protein; OSCP; Sperm flagella protein 4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
539 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P48047 | |
ATP5PO | |
Recombinant protein corresponding to Human ATP Synthase O | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only